
4-Bromo-2,6-difluorobenzotrifluoride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156243-64-0
:
Formula:C₇H₂BrF₅
MDL Number:MFCD01631616
:260.99 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together