0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
5-Bromo-2-hydroxy-3-picoline
5-Bromo-2-hydroxy-3-picoline 98%
5-Bromo-2-hydroxy-3-picoline 98%
H64253.22
:
Thermo Fisher Scientific
:
5-Bromo-2-hydroxy-3-picoline 98%
H64253.22
:
Thermo Fisher Scientific
:
Pk:
100 g
:
:
188.02 g/mol
:
170…174 °C (lit.)
:
GHS07
:
Ambient
:
89488-30-2
:
C₆H₆BrNO
:
MFCD03427657