
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103962-10-3
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD00052333
:283.04 g/mol
Melting Pt:48…55 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Related Information
Frequently Bought Together