
2-Bromo-5-formylpyridine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149806-06-4
:
Formula:C₆H₄BrNO
MDL Number:MFCD04115419
:186.01 g/mol
Melting Pt:104…110 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together