Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51298-62-5
:
Formula:C₇H₁₅N₅O₄
MDL Number:MFCD00039052
:269.69 g/mol
Melting Pt:159…161 °C
Storage Temperature:Freezer
EINECS:257-116-1
Related Information
Frequently Bought Together