Calcium benzoate hydrate, (max. 8% H₂O) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2090-05-3
:
Formula:(C₆H₅CO₂)₂Ca·xH₂O
MDL Number:MFCD00167164
:282.31 g/mol (anhy)
Storage Temperature:Ambient
EINECS:218-235-4
Related Information
Frequently Bought Together