
(R)-Piperidine-2-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1723-00-8
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD00064346
:129.16 g/mol
Melting Pt:277 °C (dec.)
EINECS:217-024-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together