Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-tert-Butyl-L-glutamate hydrate
5-tert-Butyl-L-glutamate hydrate 98%
5-tert-Butyl-L-glutamate hydrate 98%
5-tert-Butyl-L-glutamate hydrate 98%
Catalog # H63223.06
Supplier:
Thermo Fisher Scientific
CAS Number:
Expand image
5-tert-Butyl-L-glutamate hydrate 98%
Catalog # H63223.06
Supplier:
Thermo Fisher Scientific
CAS Number:
Specifications
Pk:
5 g
Specifications
About this item
Product Details
Documentation
CAS Number:
2419-56-9
:
Formula:
C₉H₁₇NO₄
MDL Number:
MFCD00038580
:
203.24 g/mol
Product Details
Documentation