
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:114676-69-6
:
Formula:C₁₁H₁₉NO₅
MDL Number:MFCD00797548
:245.27 g/mol
:GHS07
Related Information
Frequently Bought Together