



2-Adamantylzinc bromide solution 0.5 M in THF
Catalog # H58595.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:280.514 g/mol
Density:1.018
ADR:4.3,II
:Packaged under argon in resealable ChemSeal™ bottles.
Storage Temperature:Refrigerator
UN:3399
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:171860-65-4
Formula:C₁₀H₁₅BrZn
MDL Number:MFCD01311401