
2-Oxa-5-azaspiro[3.4]octane oxalate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1389264-18-9
:
Formula:C₆H₁₁NO·C₂H₂O₄
MDL Number:MFCD18325162
:203.19 g/mol
:GHS07
Related Information
Frequently Bought Together