Order Entry
Czech Republic
ContactUsLinkComponent
L-β-Proline ethyl ester hydrochloride 97%
Irritant
L-β-Proline ethyl ester hydrochloride 97%
Catalog # H57781.03
CAS Number:  
Irritant
L-β-Proline ethyl ester hydrochloride 97%
Catalog # H57781.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Formula:C₇H₁₃NO₂·HCl
MDL Number:MFCD21337911
 :179.65 g/mol
 :GHS07
Storage Temperature:Refrigerator