
6-Oxa-1-azaspiro[3.3]heptane hemioxalate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1380571-72-1
:
Formula:C₁₂H₂₀N₂O₆
MDL Number:MFCD22373634
:288.3 g/mol
:GHS07
Related Information
Frequently Bought Together