Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%
Irritant
tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%
Catalog # H57071.06
CAS Number:  
Irritant
tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%
Catalog # H57071.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

CAS Number:939793-16-5
 :
Formula:C₉H₁₆BrNO₂
 :250.14 g/mol
Melting Pt:45…53 °C
ADR:9,III
 :GHS07
MDL Number:MFCD08752478