
4-Bromo-N-(4-methoxybenzyl)aniline 97%
Catalog # H55757.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:175357-73-0
:
Formula:C₁₄H₁₄NOBr
MDL Number:MFCD00017822
:292.18 g/mol
:GHS07
Storage Temperature:Ambient