


2-Bromomethyl-6-methylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68470-59-7
:
Formula:C₇H₈BrN
MDL Number:MFCD04967229
:186.05 g/mol
Melting Pt:38…42 °C
:GHS06
:GHS05
:GHS07
Related Information
Frequently Bought Together