1,4-Bis(2-hydroxyethyl)piperazine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-96-3
:
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD00006157
:174.24 g/mol
Boiling Pt:215…220 °C (50 mmHg)
Melting Pt:133…136 °C
Storage Temperature:Ambient
EINECS:204-586-0
Related Information
Frequently Bought Together