Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,6-dimethylpyrimidine
4-Chloro-2,6-dimethylpyrimidine 97%
4-Chloro-2,6-dimethylpyrimidine 97%
4-Chloro-2,6-dimethylpyrimidine 97%
H54735.06
:
Thermo Fisher Scientific
:
4-Chloro-2,6-dimethylpyrimidine 97%
H54735.06
:
Thermo Fisher Scientific
:
Pk:
5 g
:
4472-45-1
:
:
C₆H₇ClN₂
:
MFCD00234197
:
142.03 g/mol
:
GHS07
:
Ambient