Order Entry
Czech Republic
ContactUsLinkComponent
 
Flammable
Corrosive
2,6-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-methyltetrahydrofuran)
Catalog # H54189.AE
CAS Number:  
Flammable
Corrosive
2,6-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-methyltetrahydrofuran)
Catalog # H54189.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

CAS Number:107549-26-8
 :
Formula:C₇H₅BrF₂Mg
MDL Number:MFCD11113508
 :231.32 g/mol
UN:2924
ADR:3,II
 :GHS02
 :GHS05
Storage Temperature:Ambient