
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:306960-77-0
:
Formula:C₆H₃F₃N₂O₂
MDL Number:MFCD09054851
:192.1 g/mol
Melting Pt:170…175 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together