Order Entry
Czech Republic
ContactUsLinkComponent
2-[Di(methylthio)methylidene]malononitrile 97%
Toxic
Irritant
2-[Di(methylthio)methylidene]malononitrile 97%
Catalog # H53503.14
CAS Number:  
Toxic
Irritant
2-[Di(methylthio)methylidene]malononitrile 97%
Catalog # H53503.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :170.26 g/mol
Melting Pt:79…82 °C
Storage Temperature:Ambient
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:5147-80-8
Formula:C₆H₆N₂S₂
MDL Number:MFCD00052730