

2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7223-50-9
:
Formula:C₁₁H₇NO₂
MDL Number:MFCD00065028
:185.18 g/mol
Melting Pt:148…152 °C
EINECS:230-621-4
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together