
Methyl-5-hexynoate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77758-51-1
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00671366
:126.16 g/mol
Boiling Pt:60…62 °C (10 mmHg)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together