Order Entry
Czech Republic
ContactUsLinkComponent
 
N-Boc-2-methyl-D-serine methyl ester 97%
Catalog # H52421.03
CAS Number:  
N-Boc-2-methyl-D-serine methyl ester 97%
Catalog # H52421.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.26 g/mol
Storage Temperature:Ambient
CAS Number:188476-33-7
Formula:C₁₀H₁₉NO₅
MDL Number:MFCD06797553