Order Entry
Czech Republic
ContactUsLinkComponent
N-Benzoyl-2'-deoxyadenosine 98+%
N-Benzoyl-2'-deoxyadenosine 98+%
Catalog # H52303.06
CAS Number:  
N-Benzoyl-2'-deoxyadenosine 98+%
Catalog # H52303.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:4546-72-9
 :
Formula:C₁₇H₁₇N₅O₄
MDL Number:MFCD00009628
 :355.35 g/mol
Storage Temperature:Ambient
EINECS:224-903-6
 :MFCD00149575