2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:196207-58-6
:
Formula:C₄₁H₆₄B₂O₄
MDL Number:MFCD16294554
:642.58 g/mol
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Frequently Bought Together