
Pyridine-2-carboxamidoxime 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1772-01-6
:
Formula:C₆H₇N₃O
MDL Number:MFCD00085159
:137.14 g/mol
Melting Pt:115…120 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together