Order Entry
Czech Republic
ContactUsLinkComponent
1-Pyridin-2-yl[1,4]diazepane ≥98%
1-Pyridin-2-yl[1,4]diazepane ≥98%
Catalog # H50946.MD
CAS Number:  
1-Pyridin-2-yl[1,4]diazepane ≥98%
Catalog # H50946.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :177.25 g/mol
ADR:,
CAS Number:287114-32-3
Formula:C₁₀H₁₅N₃
MDL Number:MFCD06409204