Order Entry
Czech Republic
ContactUsLinkComponent
4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
Irritant
4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
Catalog # H50804.03
CAS Number:  
Irritant
4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
Catalog # H50804.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:902836-38-8
 :
Formula:C₁₄H₁₆ClN₃
MDL Number:MFCD08060997
 :261.76 g/mol
 :GHS07
Storage Temperature:Ambient