
3-(3-Methoxyphenyl)-1H-pyrazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:144026-74-4
:
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD01940432
:174.2 g/mol
Melting Pt:90…91 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together