
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethanamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62821-88-9
:
Formula:C₇H₁₃N₃
MDL Number:MFCD04967722
:139.2 g/mol
ADR:8,III
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together