
N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6342-21-8
:
Formula:C₁₀H₁₆N₂
MDL Number:MFCD01631934
:164.25 g/mol
ADR:8,III
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together