Order Entry
Czech Republic
ContactUsLinkComponent
6-Pyrrolidin-1-ylnicotinaldehyde 98%
Irritant
6-Pyrrolidin-1-ylnicotinaldehyde 98%
Catalog # H50126.03
CAS Number:  
Irritant
6-Pyrrolidin-1-ylnicotinaldehyde 98%
Catalog # H50126.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:261715-39-3
 :
Formula:C₁₀H₁₂N₂O
MDL Number:MFCD06200881
 :176.22 g/mol
 :GHS07
Storage Temperature:Ambient