1,3-Di-tert-butyl-2-methoxybenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1516-95-6
:
Formula:C₁₅H₂₄O
MDL Number:MFCD20483412
:220.356 g/mol
Boiling Pt:124 °C (9 mmHg)
Storage Temperature:Ambient
Related Information
Frequently Bought Together