Palladium : Platinum (4:1) on carbon, paste E101023-4/1
Catalog # H37009.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd:Pt
MDL Number:MFCD03613605