Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
2-Methoxybenzenesulphonyl chloride 97%
Catalog # H34455.03
CAS Number:  
Corrosive
2-Methoxybenzenesulphonyl chloride 97%
Catalog # H34455.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:10130-87-7
 :
Formula:C₇H₇ClO₃S
MDL Number:MFCD01961367
 :206.65 g/mol
Melting Pt:50…54 °C
Storage Temperature:Ambient
ADR:8,III
 :GHS05