
Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1027511-41-6
:
Formula:C₉H₈BrN₃O₂
MDL Number:MFCD10574908
:270.08 g/mol
Melting Pt:144…146 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together