
3-(3-Chloro-4-fluorophenyl)propionic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:881190-93-8
:
Formula:C₉H₈ClFO₂
MDL Number:MFCD04116056
:202.61 g/mol
Melting Pt:56…59 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together