
Palladium 5% on activated carbon paste (50% water) 5R437
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
UN:1325
ADR:4.1,III
:GHS02
Frequently Bought Together