
4,6-Dichloro-2-(trifluoromethyl)pyrimidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:705-24-8
:
Formula:C₅HCl₂F₃N₂
MDL Number:MFCD08436597
:216.98 g/mol
Boiling Pt:50…52 °C (5 mmHg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together