Order Entry
Czech Republic
ContactUsLinkComponent
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol 96%
Irritant
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol 96%
Catalog # H33012.ME
CAS Number:  
Irritant
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol 96%
Catalog # H33012.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
Melting Pt:194…195 °C
 :247.34 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:500112-74-3
Formula:C₁₁H₉N₃S₂
MDL Number:MFCD06254379