
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108354-78-5
:
Formula:C₁₀H₁₃NO₂S
MDL Number:MFCD00130099
:211.28 g/mol
Melting Pt:126…129 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together