
2-Fluoro-4-(trifluoromethyl)nicotinic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1040681-74-0
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD10687403
:209.10 g/mol
Melting Pt:91…93 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together