
2-Furaldehyde-2,2-dimethylhydrazone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14064-21-2
:
Formula:C₇H₁₀N₂O
MDL Number:MFCD00010685
:138.17 g/mol
Boiling Pt:69 °C (2 mmHg)
Density:1.048
Flash Pt:92 °C (198 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together