

2,2-Difluoropropionic acid ethyl ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28781-85-3
:
Formula:C₅H₈F₂O₂
MDL Number:MFCD06657970
:138.12 g/mol
Boiling Pt:24 °C (20 mmHg)
Density:1.102
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together