4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149777-83-3
:
Formula:C₁₅H₂₁BO₃
MDL Number:MFCD04038966
:260.14 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together