
1,1'-Thiocarbonyldi-2(1H)-pyridone 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102368-13-8
:
Formula:C₁₁H₈N₂O₂S
MDL Number:MFCD00075238
:232.26 g/mol
Melting Pt:163…166 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together