
tert-Butyl-3-hydroxyazetidine-1-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141699-55-0
:
Formula:C₈H₁₅NO₃
MDL Number:MFCD04115305
Melting Pt:40…47 °C
:173.21 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together