ContactUsLinkComponent
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
  H27830.06
 :  
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
  H27830.06
 :  
  • Pk:
    5 g
  • Seq:
    3173
 :845-46-5
 :
 :C₁₅H₁₄N₃O₂Na
 :MFCD00020350
 :291.28 g/mol
 :>300 °C
 :GHS07
 :Ambient