
Ethyl-2-hydroxy-6-methylpyridine-4-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:150190-03-7
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD09953494
:181.20 g/mol
Melting Pt:188…190 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together